Assay Details
| Gene Symbol: UCHL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305615.1 | NP_001292544.1 | 6 - 7 | 695 | 73 | |
| NR_131183.1 | 6 - 7 | 695 | 73 | |||
| XM_025152804.1 | XP_025008572.1 | 5 - 6 | 476 | 73 | ||
| XM_025152805.1 | XP_025008573.1 | 4 - 5 | 415 | 73 | ||
| XM_025152806.1 | XP_025008574.1 | 5 - 6 | 470 | 73 | ||
| GenBank mRNA | AJ720133.1 | - | 6 - 7 | 695 | 73 | |
More Information
Set Membership: |
|

UCHL5
Genomic Map