Assay Details
| Gene Symbol: CDC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006536.1 | NP_001006536.1 | 10 - 11 | 1380 | 65 | |
| GenBank mRNA | AJ720948.1 | - | 10 - 11 | 1380 | 65 | |
More Information
Set Membership: |
|

CDC20
Genomic Map