Assay Details
| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006545.1 | NP_001006545.1 | 3 - 4 | 881 | 84 | |
| GenBank mRNA | AJ720003.1 | - | 3 - 4 | 881 | 84 | |
More Information
Set Membership: |
|

TBCCD1
Genomic Map