Assay Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006553.1 | NP_001006553.1 | 5 - 6 | 453 | 119 | |
| GenBank mRNA | AJ720223.1 | - | 5 - 6 | 453 | 119 | |
More Information
Set Membership: |
|

NDUFB5
Genomic Map