Assay Details
| Gene Symbol: ANKRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006565.1 | NP_001006565.1 | 3 - 4 | 461 | 70 | |
| XM_015277286.2 | XP_015132772.1 | 2 - 3 | 696 | 70 | ||
| XM_015277287.2 | XP_015132773.1 | 2 - 3 | 454 | 70 | ||
| GenBank mRNA | AJ720862.1 | - | 3 - 4 | 461 | 70 | |
More Information
Set Membership: |
|

ANKRA2
Genomic Map