Assay Details
| Gene Symbol: DDX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006568.1 | NP_001006568.1 | 9 - 10 | 1045 | 119 | |
| XM_015296933.2 | XP_015152419.1 | 9 - 10 | 1120 | 119 | ||
| XM_025142541.1 | XP_024998309.1 | 9 - 10 | 1100 | 119 | ||
| GenBank mRNA | AJ719463.1 | - | 9 - 10 | 1045 | 119 | |
| BX936036.1 | - | 9 - 10 | 1062 | 119 | ||
More Information
Set Membership: |
|

DDX19
Genomic Map