Assay Details
| Gene Symbol: GOLGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006570.1 | NP_001006570.1 | 3 - 4 | 550 | 73 | |
| GenBank mRNA | AJ719805.1 | - | 3 - 4 | 550 | 73 | |
| CR387916.1 | - | 3 - 4 | 436 | 73 | ||
More Information
Set Membership: |
|

GOLGA7
Genomic Map