Assay Details
| Gene Symbol: DAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007473.1 | NP_001007474.1 | 1 - 2 | 249 | 100 | |
| GenBank mRNA | AF221078.1 | - | 1 - 2 | 211 | 100 | |
| AF221079.1 | - | 1 - 2 | 211 | 100 | ||
| U83627.1 | - | 1 - 2 | 249 | 100 | ||
More Information
Set Membership: |
|

DAD1
Genomic Map