Assay Details
| Gene Symbol: PRRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277724.2 | NP_001264653.1 | 2 - 3 | 655 | 77 | |
| XM_015290230.2 | XP_015145716.1 | 2 - 3 | 742 | 77 | ||
| XM_015290231.2 | XP_015145717.1 | 2 - 3 | 741 | 77 | ||
| GenBank mRNA | AF461698.1 | - | 2 - 3 | 471 | 77 | |
| BU311875.1 | - | 2 - 3 | 242 | 77 | ||
| D13433.1 | - | 2 - 3 | 652 | 77 | ||
| S69088.1 | - | 2 - 3 | 450 | 77 | ||
More Information
Set Membership: |
|

PRRX1
Genomic Map