Assay Details
| Gene Symbol: TANGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007836.1 | NP_001007837.1 | 6 - 7 | 563 | 137 | |
| XM_015275249.2 | XP_015130735.1 | 6 - 7 | 563 | 137 | ||
| XM_015275250.2 | XP_015130736.1 | 6 - 7 | 639 | 137 | ||
| XM_015275251.2 | XP_015130737.1 | 6 - 7 | 690 | 137 | ||
| XM_015275252.2 | XP_015130738.1 | 6 - 7 | 771 | 137 | ||
| XM_015275253.2 | XP_015130739.1 | 6 - 7 | 703 | 137 | ||
| XM_025155442.1 | XP_025011210.1 | 6 - 7 | 1695 | 137 | ||
| GenBank mRNA | AJ720940.1 | - | 6 - 7 | 563 | 137 | |
More Information
Set Membership: |
|

TANGO2
Genomic Map