Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007837.1 | NP_001007838.1 | 4 - 5 | 473 | 93 | |
| XM_015275294.2 | XP_015130780.1 | 4 - 5 | 789 | 93 | ||
| XM_015275295.2 | XP_015130781.1 | 4 - 5 | 780 | 93 | ||
| XM_015275296.2 | XP_015130782.1 | 3 - 4 | 483 | 93 | ||
| GenBank mRNA | AJ720772.1 | - | 4 - 5 | 473 | 93 | |
| CR352820.1 | - | 2 - 3 | 688 | 93 | ||
More Information
Set Membership: |
|

VPS29
Genomic Map