Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007838.1 | NP_001007839.1 | 2 - 3 | 209 | 77 | |
| XM_025155502.1 | XP_025011270.1 | 2 - 3 | 215 | 77 | ||
| GenBank mRNA | AJ719876.1 | - | 2 - 3 | 209 | 77 | |
| CR388618.1 | - | 2 - 3 | 191 | 77 | ||
More Information
Set Membership: |
|

SNRPD3
Genomic Map