Assay Details
| Gene Symbol: CBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007848.1 | NP_001007849.1 | 4 - 5 | 638 | 101 | |
| XM_015275711.2 | XP_015131197.1 | 4 - 5 | 721 | 101 | ||
| GenBank mRNA | AJ720990.1 | - | 4 - 5 | 638 | 101 | |
| BX931148.1 | - | 4 - 5 | 552 | 101 | ||
More Information
Set Membership: |
|

CBLL1
Genomic Map