Assay Details
| Gene Symbol: RASSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007851.1 | NP_001007852.1 | 3 - 4 | 595 | 79 | |
| XM_015281587.2 | XP_015137073.1 | 3 - 4 | 2415 | 79 | ||
| XM_015281591.1 | XP_015137077.1 | 4 - 5 | 810 | 79 | ||
| XM_015281595.2 | XP_015137081.1 | 3 - 4 | 497 | 79 | ||
| GenBank mRNA | AJ720647.1 | - | 3 - 4 | 595 | 79 | |
More Information
Set Membership: |
|

RASSF3
Genomic Map