Assay Details
| Gene Symbol: GID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007871.1 | NP_001007872.1 | 3 - 4 | 353 | 84 | |
| GenBank mRNA | AJ720027.1 | - | 3 - 4 | 353 | 84 | |
| BX929274.1 | - | 3 - 4 | 364 | 84 | ||
More Information
Set Membership: |
|

GID8
Genomic Map