Assay Details
| Gene Symbol: SPATA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007873.1 | NP_001007874.1 | 2 - 3 | 484 | 118 | |
| XM_015296412.2 | XP_015151898.1 | 2 - 3 | 746 | 118 | ||
| GenBank mRNA | AJ720887.1 | - | 2 - 3 | 484 | 118 | |
More Information
Set Membership: |
|

SPATA2
Genomic Map