Assay Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007905.2 | NP_001007906.1 | 1 - 2 | 383 | 54 | |
| GenBank mRNA | BX933632.1 | - | 1 - 2 | 119 | 54 | |
| BX933635.1 | - | 1 - 2 | 136 | 54 | ||
| BX934589.1 | - | 1 - 2 | 138 | 54 | ||
| BX935045.2 | - | 1 - 2 | 125 | 54 | ||
| BX935881.1 | - | 1 - 2 | 123 | 54 | ||
| BX935913.1 | - | 1 - 2 | 379 | 54 | ||
| BX950755.2 | - | 1 - 2 | 117 | 54 | ||
More Information
Set Membership: |
|

PSMB1
Genomic Map