Assay Details
| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007917.2 | NP_001007918.1 | 6 - 7 | 827 | 59 | |
| GenBank mRNA | AJ720634.1 | - | 6 - 7 | 792 | 59 | |
More Information
Set Membership: |
|

MPP1
Genomic Map