Assay Details
| Gene Symbol: FABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007923.1 | NP_001007924.1 | 2 - 3 | 238 | 81 | |
| GenBank mRNA | BX930518.1 | - | 2 - 3 | 261 | 81 | |
| BX932340.2 | - | 2 - 3 | 255 | 81 | ||
| BX933597.1 | - | 2 - 3 | 258 | 81 | ||
More Information
Set Membership: |
|

FABP2
Genomic Map