Assay Details
| Gene Symbol: RNF141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007925.1 | NP_001007926.1 | 3 - 4 | 394 | 84 | |
| XM_015286375.2 | XP_015141861.1 | 3 - 4 | 360 | 84 | ||
| XM_025150670.1 | XP_025006438.1 | 3 - 4 | 331 | 84 | ||
| GenBank mRNA | AJ719510.1 | - | 3 - 4 | 394 | 84 | |
More Information
Set Membership: |
|

RNF141
Genomic Map