Assay Details
| Gene Symbol: LEPROT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007958.1 | NP_001007959.1 | 2 - 3 | 171 | 122 | |
| XM_015291028.2 | XP_015146514.1 | 2 - 3 | 173 | 122 | ||
| GenBank mRNA | AJ720495.1 | - | 2 - 3 | 171 | 122 | |
More Information
Set Membership: |
|

LEPROT
Genomic Map