Assay Details
| Gene Symbol: LEPROT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007958.1 | NP_001007959.1 | 3 - 4 | 357 | 150 | |
| GenBank mRNA | AJ720495.1 | - | 3 - 4 | 357 | 150 | |
More Information
Set Membership: |
|

LEPROT
Genomic Map