Assay Details
| Gene Symbol: ENSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007965.1 | NP_001007966.1 | 1 - 2 | 110 | 61 | |
| GenBank mRNA | AJ720826.1 | - | 1 - 2 | 110 | 61 | |
More Information
Set Membership: |
|

ENSA
Genomic Map