Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007967.1 | NP_001007968.1 | 2 - 3 | 168 | 65 | |
| XM_015282954.2 | XP_015138440.1 | 3 - 4 | 238 | 65 | ||
More Information
Set Membership: |
|

RPL30
Genomic Map