Assay Details
| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007970.1 | NP_001007971.1 | 5 - 6 | 484 | 79 | |
| XM_015301808.2 | XP_015157294.1 | 5 - 6 | 622 | 79 | ||
| GenBank mRNA | AJ719969.1 | - | 5 - 6 | 484 | 79 | |
More Information
Set Membership: |
|

ARL2BP
Genomic Map