Assay Details
| Gene Symbol: ACTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008446.1 | NP_001008446.1 | 2 - 3 | 627 | 71 | |
| GenBank mRNA | AJ720530.1 | - | 2 - 3 | 627 | 71 | |
More Information
Set Membership: |
|

ACTR5
Genomic Map