Assay Details
| Gene Symbol: LGALSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008456.1 | NP_001008456.1 | 3 - 4 | 320 | 81 | |
| GenBank mRNA | AJ721082.1 | - | 3 - 4 | 320 | 81 | |
| CR353568.1 | - | 3 - 4 | 184 | 81 | ||
More Information
Set Membership: |
|

LGALSL
Genomic Map