Assay Details
| Gene Symbol: BRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008458.1 | NP_001008458.1 | 9 - 10 | 1026 | 62 | |
| XM_015283708.2 | XP_015139194.1 | 9 - 10 | 62 | |||
| XM_015283709.2 | XP_015139195.1 | 9 - 10 | 62 | |||
| GenBank mRNA | AJ719374.1 | - | 9 - 10 | 1026 | 62 | |
| CR389763.1 | - | 6 - 7 | 62 | |||
More Information
Set Membership: |
|

BRE
Genomic Map