Assay Details
| Gene Symbol: PGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008463.1 | NP_001008463.1 | 2 - 3 | 325 | 65 | |
| GenBank mRNA | AJ721095.1 | - | 2 - 3 | 325 | 65 | |
More Information
Set Membership: |
|

PGS1
Genomic Map