Assay Details
| Gene Symbol: DR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008478.1 | NP_001008478.1 | 1 - 2 | 615 | 99 | |
| GenBank mRNA | AJ719281.1 | - | 1 - 2 | 615 | 99 | |
| BX931827.2 | - | 1 - 2 | 310 | 99 | ||
More Information
Set Membership: |
|

DR1
Genomic Map