Assay Details
| Gene Symbol: RP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008680.1 | NP_001008680.1 | 3 - 4 | 1059 | 69 | |
| GenBank mRNA | AJ721100.1 | - | 3 - 4 | 1059 | 69 | |
More Information
Set Membership: |
|

RP2
Genomic Map