Assay Details
| Gene Symbol: SUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204135.2 | NP_989466.1 | 2 - 3 | 171 | 121 | |
| XM_015289365.2 | XP_015144851.1 | 2 - 3 | 181 | 121 | ||
| GenBank mRNA | AF461015.1 | - | 2 - 3 | 179 | 121 | |
| AJ719470.1 | - | 2 - 3 | 171 | 121 | ||
More Information
Set Membership: |
|

SUMO1
Genomic Map