Assay Details
| Gene Symbol: SOX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204731.1 | NP_990062.1 | 2 - 3 | 684 | 126 | |
| GenBank mRNA | AF228664.1 | - | 2 - 3 | 684 | 126 | |
More Information
Set Membership: |
|

SOX8
Genomic Map