Assay Details
| Gene Symbol: CTAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204732.1 | NP_990063.1 | 3 - 4 | 382 | 97 | |
| XM_025148519.1 | XP_025004287.1 | 5 - 6 | 4035 | 97 | ||
| GenBank mRNA | AF233518.1 | - | 3 - 4 | 382 | 97 | |
More Information
Set Membership: |
|

CTAGE1
Genomic Map