Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204756.1 | NP_990087.1 | 2 - 3 | 228 | 96 | |
| XM_015294133.1 | XP_015149619.1 | 2 - 3 | 251 | 96 | ||
| XM_015294134.2 | XP_015149620.1 | 2 - 3 | 186 | 96 | ||
| GenBank mRNA | D16187.1 | - | 2 - 3 | 228 | 96 | |
More Information
Set Membership: |
|

MAFK
Genomic Map