Assay Details
| Gene Symbol: VSX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204768.1 | NP_990099.1 | 3 - 4 | 644 | 71 | |
| XM_025150559.1 | XP_025006327.1 | 3 - 4 | 933 | 71 | ||
| GenBank mRNA | AF178671.1 | - | 3 - 4 | 644 | 71 | |
More Information
Set Membership: |
|

VSX2
Genomic Map