Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204778.1 | NP_990109.1 | 11 - 12 | 1327 | 96 | |
| XM_015279900.2 | XP_015135386.1 | 12 - 13 | 1507 | 96 | ||
| XM_015279904.2 | XP_015135390.1 | 12 - 13 | 1501 | 96 | ||
| XM_015279909.2 | XP_015135395.1 | 12 - 13 | 1434 | 96 | ||
| GenBank mRNA | AF166094.1 | - | 11 - 12 | 1327 | 96 | |
More Information
Set Membership: |
|

MRE11
Genomic Map