Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204778.1 | NP_990109.1 | 18 - 19 | 2062 | 74 | |
| XM_015279900.2 | XP_015135386.1 | 19 - 20 | 2242 | 74 | ||
| XM_015279904.2 | XP_015135390.1 | 19 - 20 | 2236 | 74 | ||
| XM_015279909.2 | XP_015135395.1 | 19 - 20 | 2169 | 74 | ||
| GenBank mRNA | AF166094.1 | - | 18 - 19 | 2062 | 74 | |
More Information
Set Membership: |
|

MRE11
Genomic Map