Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204778.1 | NP_990109.1 | 8 - 9 | 1018 | 68 | |
| XM_015279900.2 | XP_015135386.1 | 9 - 10 | 1198 | 68 | ||
| XM_015279904.2 | XP_015135390.1 | 9 - 10 | 1192 | 68 | ||
| XM_015279909.2 | XP_015135395.1 | 9 - 10 | 1125 | 68 | ||
| GenBank mRNA | AF166094.1 | - | 8 - 9 | 1018 | 68 | |
More Information
Set Membership: |
|

MRE11
Genomic Map