Assay Details
| Gene Symbol: TUBA8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205444.1 | NP_990775.1 | 2 - 3 | 379 | 95 | |
| GenBank mRNA | M16030.1 | - | 2 - 3 | 379 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBA8B| Gene Symbol: TUBA8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205444.1 | NP_990775.1 | 2 - 3 | 379 | 95 | |
| GenBank mRNA | M16030.1 | - | 2 - 3 | 379 | 95 | |
Set Membership: |
|