Assay Details
| Gene Symbol: THRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252221.2 | NP_001239150.1 | 3 - 4 | 600 | 84 | |
| NM_205447.2 | NP_990778.2 | 3 - 4 | 318 | 84 | ||
| XM_015281323.2 | XP_015136809.1 | 5 - 6 | 2019 | 84 | ||
| XM_015281324.2 | XP_015136810.1 | 5 - 6 | 427 | 84 | ||
| XM_015281325.2 | XP_015136811.1 | 5 - 6 | 467 | 84 | ||
| XM_015281326.2 | XP_015136812.1 | 4 - 5 | 340 | 84 | ||
| XM_025147069.1 | XP_025002837.1 | 4 - 5 | 437 | 84 | ||
| XM_025147070.1 | XP_025002838.1 | 5 - 6 | 461 | 84 | ||
| XM_025147071.1 | XP_025002839.1 | 4 - 5 | 428 | 84 | ||
| XM_025147072.1 | XP_025002840.1 | 5 - 6 | 458 | 84 | ||
| XM_025147073.1 | XP_025002841.1 | 4 - 5 | 337 | 84 | ||
| XM_025147074.1 | XP_025002842.1 | 4 - 5 | 436 | 84 | ||
| GenBank mRNA | M65207.1 | - | 487 | 84 | ||
| X17504.1 | - | 341 | 84 | |||
| Z50188.1 | - | 296 | 84 | |||
More Information
Set Membership: |
|

THRB
Genomic Map