Assay Details
| Gene Symbol: SNAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205458.1 | NP_990789.1 | 6 - 7 | 554 | 76 | |
| XM_015283456.2 | XP_015138942.1 | 7 - 8 | 757 | 76 | ||
| XM_015283457.2 | XP_015138943.1 | 7 - 8 | 757 | 76 | ||
| GenBank mRNA | M57957.1 | - | 6 - 7 | 554 | 76 | |
More Information
Set Membership: |
|

SNAP25
Genomic Map