Assay Details
| Gene Symbol: CYP2C23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001757.1 | NP_001001757.1 | 4 - 5 | 697 | 155 | |
| GenBank mRNA | M25469.1 | - | 4 - 5 | 697 | 155 | |
More Information
Set Membership: |
|

CYP2C23b
Genomic Map