Assay Details
| Gene Symbol: RPL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204152.1 | NP_989483.1 | 4 - 5 | 529 | 136 | |
| GenBank mRNA | AY032864.1 | - | 4 - 5 | 529 | 136 | |
More Information
Set Membership: |
|

RPL6
Genomic Map