Assay Details
| Gene Symbol: CYP2C23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001616.1 | NP_001001616.1 | 9 | 2650 | 138 | |
| GenBank mRNA | M13454.1 | - | 9 | 2650 | 138 | |
More Information
Set Membership: |
|

CYP2C23a
Genomic Map