Assay Details
| Gene Symbol: HBBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004390.2 | NP_001004390.1 | 2 | 253 | 131 | |
| GenBank mRNA | CD762024.1 | - | 2 | 288 | 131 | |
| V00412.1 | - | 253 | 131 | |||
More Information
Set Membership: |
|

HBBR
Genomic Map