Assay Details
| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031489.2 | NP_001026660.1 | 3 | 433 | 102 | |
| GenBank mRNA | BX265880.3 | - | 3 | 441 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HBE1| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031489.2 | NP_001026660.1 | 3 | 433 | 102 | |
| GenBank mRNA | BX265880.3 | - | 3 | 441 | 102 | |
Set Membership: |
|