Assay Details
| Gene Symbol: APOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205525.4 | NP_990856.1 | 4 | 750 | 149 | |
| XM_015297971.2 | XP_015153457.1 | 5 | 657 | 149 | ||
| GenBank mRNA | CR352616.1 | - | 5 | 633 | 149 | |
| CR385590.1 | - | 4 | 752 | 149 | ||
| CR385797.1 | - | 4 | 737 | 149 | ||
| CR386713.1 | - | 5 | 646 | 149 | ||
| CR390380.1 | - | 3 | 441 | 149 | ||
| CR523874.1 | - | 5 | 613 | 149 | ||
| M18746.1 | - | 4 | 750 | 149 | ||
| M25559.1 | - | 3 | 722 | 149 | ||
More Information
Set Membership: |
|

APOA1
Genomic Map