Assay Details
| Gene Symbol: DNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206957.1 | NP_996840.1 | 9 | 941 | 124 | |
| GenBank mRNA | AB013751.1 | - | 9 | 928 | 125 | |
| AJ131751.1 | - | 9 | 941 | 124 | ||
More Information
Set Membership: |
|

DNASE1
Genomic Map