Assay Details
| Gene Symbol: IFNL3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128496.1 | NP_001121968.1 | 1 - 2 | 163 | 91 | |
| GenBank mRNA | AM773754.1 | - | 1 - 2 | 163 | 91 | |
| EF587763.1 | - | 1 - 2 | 163 | 91 | ||
| EU399904.1 | - | 1 - 2 | 163 | 91 | ||
| Gene Symbol: IFNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289181.2 | XP_015144667.1 | 1 - 2 | 1108 | 91 | |
More Information
Set Membership: |
|

IFNL3A
Genomic Map